Would you like to log in again?
VIAL AUTOSAMPLER SNAP TOP AMBER GL 2ML
Supplier: MicroSolv Technology Corp
Easy Purchase Pack Basik* Autosampler vial 2ml^ Easy Purchase Pack Basik* Autosampler vial, snap top, Material: amber glass, with write on patch, 9-425mm threads, and blue screw caps with fitted single slit Soft-Guard silicone/PTFE septa, BASIK brand, Dimension: 12x32mm, Size: 2ml
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VIAL AUTOSAMPLER SNAP TOP AMBER GL 2ML 76047-160 9502C-E-3WAP Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty