Would you like to log in again?
Plain Toe PVC Safety Boots with Steel Shank, Onguard
Supplier: Onguard
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
81011-12 81011-11 81011-08 81011-07 81011-09 81011-06 81011-10
10466-476PR 45.22 USD InStock
10466-476 76047-240 76047-216 76047-238 76047-214 76047-236 76047-212
Plain Toe PVC Safety Boots with Steel Shank, Onguard
Shoes and Boots
Boots provide reliable, industry proven protection.
  • Comfortable, waterproof protection
  • Superior traction on slippery surfaces
  • PVC upper provides mild chemical resistance and helps comply with HACCP guidelines
Injection-molded seamless construction with replaceable cushion insoles. Boots feature a Safety-Loc™ sole, cut-off bands for height adjustment, steel shank, and pull-up/kick-off lugs for easy on/off. These boots offer a perfect fit, optimizing comfort for everyday use. They are 100% waterproof to keep feet dry under wet and dirty conditions, and offer protection against a wide variety of chemicals. Excellent for use in commercial fishing, shrimping, and food processing.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty