Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® TraceableLIVE® Wi-Fi Data Logging Hygrometer and Thermometers with Remote Notification
Supplier: Avantor
VWR®
61010-48 61010-49
76047-260EA 611.02 USD InStock
76047-260 76047-258
VWR® TraceableLIVE® Wi-Fi Data Logging Hygrometer and Thermometers with Remote Notification
Data Loggers
VWR® TraceableLIVE® is the simple, efficient, and reliable way to stay connected to critical environments, wherever you go.
  • Monitor temperature and humidity readings locally and remotely by connecting to existing Wi-Fi
  • Alerts: humidity alarm, temperature alarm, connectivity interruption, low battery
  • Cloud-based data interface with no local software required; web, Android, Apple IOS data visibility
  • Continuous data transmission and monitoring using SSL encrypted SECURE connection
  • Receive Mobile Push, Email, or Text notifications
  • Unlimited scalability; use TraceableLIVE® in individual labs or an entire network of facilities
  • Ambient probe and cable probe units available
  • Multiple point humidity and temperature calibration, traceable to NIST
With TraceableLIVE®, stay connected to the lab with a smartphone, tablet, or personal computer. By simply using the TraceableLIVE® app, or visiting Traceablelive.com on any browser, users can view current humidity and temperatures, control alarm parameters, view datalogging history, and view and generate reports. TraceableLIVE® Standard and Premium subscription plans are now available on vwr.com (https://us.vwr.com/store/product/35955573/traceablelive-subscription-coupon-codes-cole-parmer).
Certifications: CE marked. RoHS compliant. Traceable to NIST for accuracy. Multi-point calibration on an individually-numbered Traceable® certificate which assures accuracy from an ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM mutual recognition agreement.
Ordering information: Units are supplied with probe, quick start guide, batteries, wall mount, Velcro®, magnetic strips, and an ISO 17025 Traceable® certificate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty