Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® TraceableLIVE® Wi-Fi Data Logging LN₂ Thermometers with Remote Notification
Supplier: VWR
VWR®
61010-67 61010-76
76047-270EA 910.72 USD InStock
76047-270 76047-268
VWR® TraceableLIVE® Wi-Fi Data Logging LN₂ Thermometers with Remote Notification
Data Loggers
Covering cryo needs for sensitive, irreplaceable bioligicals.
  • Monitor temperature readings locally and remotely by connecting to existing Wi-Fi
  • Ideal for LN₂ storage vessels, 2 probe version permits monitoring the LN₂ dewar and ambient air temperature simultaneously
  • Alerts: temperature alarm, connectivity interruption, low battery
  • Cloud-based data interface with no local software required; web, Android, Apple IOS data visibility
  • Continuous data transmission and monitoring using SSL encrypted SECURE connection
  • Receive Mobile Push, Email, or Text notifications
  • Unlimited scalability; use TraceableLIVE® in individual labs or an entire network of facilities
  • Multiple point temperature calibration traceable to NIST, with calibration points of –196°C, –80°C, 0°C, and 100°C
Platinum sensor, stainless-steel probe has fast-response platinum RTD sensor, stainless-steel 316 probe displays the slightest temperature change instantly. Place probe inside dewar-readings are updated three times a second. Probe has a Ø of 0.12", stem length of 6" and a cable length of 10'.
VWR® TraceableLIVE® is the simple, efficient, and reliable way to stay connected to critical environments, wherever you go. Audible and visual alarms helps those present in the lab identify any out-of-range conditions. High-accuracy thermometer, high-impact, chemical-resistant ABS plastic case. Ideal for monitoring long term sensitive biologicals, stem cell and more., that are irreplaceable. Works with any liquid nitrogen/cryogenic storage, dewar.
With TraceableLIVE®, stay connected to the lab with a smartphone, tablet, or personal computer. By simply using the TraceableLIVE® app, or visiting Traceablelive.com on any browser, users can view current humidity and temperatures, control alarm parameters, view datalogging history, and view and generate reports. TraceableLIVE® Standard and Premium subscription plans are now available on website (https://us.vwr.com/store/product/35955573/traceablelive-subscription-coupon-codes-cole-parmer).
Certifications: CE marked. RoHS compliant. Traceable to NIST for accuracy. Multi-point calibration on an individually-numbered Traceable® certificate which assures accuracy from an ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM mutual recognition agreement.
Ordering information: Units are supplied with two probe, quick start guide, batteries, wall mount, Velcro®, magnetic strips, and an ISO 17025 Traceable® certificate.
Order Now
ORDER
SPECIFICATIONS
SERVICES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty