Would you like to log in again?
VWR® TraceableLIVE® Wi-Fi Datalogging Hi-Temp Thermometers with Remote Notification
Supplier: VWR
VWR®
61011-50 61011-48
76047-274EA 802.58 USD InStock
76047-274 76047-272
VWR® TraceableLIVE® Wi-Fi Datalogging Hi-Temp Thermometers with Remote Notification
Data Loggers
VWR® TraceableLIVE® is the simple, efficient, and reliable way to stay connected to critical environments, wherever you go.
  • Monitor temperature readings locally and remotely by connecting to existing Wi-Fi
  • Ideal for incubators, ovens, and other high temperature applications (both probe and cable can be exposed to temperatures up to 300°C on a continuous basis)
  • Alerts: temperature alarm, connectivity interruption, low battery
  • Cloud-based data interface with no local software required; web, Android, Apple IOS data visibility
  • Continuous data transmission and monitoring using SSL encrypted SECURE connection
  • Receive Mobile Push, Email, or Text notifications
  • Unlimited scalability; use TraceableLIVE® in individual labs or an entire network of facilities
  • Multiple point temperature calibration traceable to NIST, with calibration points of 0 °C, 100 °C, 200 °C, and 2900 °C
Audible and visual alarms helps those present in the lab identify any out-of-range conditions. Heat-resistant cable and probe. Fast-response Platinum RTD sensor, stainless-steel 316 probe displays the slightest temperature change instantly. Place probe inside oven-readings are updated three times a second. Probe has a Ø of 0.12", stem length of 6" and a cable length of 10'. Ideal for monitoring ovens, industrial processes, and any high-temperature application.
With TraceableLIVE®, stay connected to the lab with a smartphone, tablet, or personal computer. By simply using the TraceableLIVE® app, or visiting Traceablelive.com on any browser, users can view current humidity and temperatures, control alarm parameters, view datalogging history, and view and generate reports. TraceableLIVE® Standard and Premium subscription plans are now available on vwr.com (https://us.vwr.com/store/product/35955573/traceablelive-subscription-coupon-codes-cole-parmer).
Certifications: CE marked. RoHS compliant. Traceable to NIST for accuracy. Multi-point calibration on an individually-numbered Traceable® Certificate which assures accuracy from an ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement.
Ordering information: Units are supplied with one or two probes, Quick Start Guide, batteries, wall mount, Velcro®, hook and loop tape, magnetic strips, and an ISO 17025 Traceable® Certificate.
Order Now
ORDER
SPECIFICATIONS
SERVICES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty