Would you like to log in again?
VWR® Manual Crimping and Decapping Tools
Supplier: VWR
VWR®
76047-562A 76047-564A 76047-558A 76047-556A 76047-566A 76047-570A
76047-558APK 303.19 USD InStock
76047-558A 76047-556A 76047-566A 76047-564A 76047-562A 76047-570A
VWR® Manual Crimping and Decapping Tools
Crimpers, Cappers and Decappers
Screw-stop in crimping handle allows for adjustment of crimping pressure. Decrimping tools provide convenient method for crimp seal removal.
  • Easy and convenient handling
  • Durable, high quality construction
  • Adjustable screw setting facilitates optimum crimping
Adjustable in crimping pressure limitation by screw in handle.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty