Would you like to log in again?
VWR® Eyewash Stations
Supplier: VWR
VWR®
76049-032 76049-028 76049-030 76049-022
76049-030CS 237.47 USD InStock
76049-030 76049-032 76049-022 76049-028
VWR® Eyewash Stations
First Aid Supplies Eyewash Stations and Eyewash Bottles
VWR® eyewash stations are wall-mountable, providing quick access for emergencies.
  • Wall-mountable stations in single or double configurations
  • 16 or 32 oz. bottle options
  • Includes Physicians Care eyewash solution
The stations are configured to hold either one or two bottles, available in 16 oz.
PhysiciansCare sterile isotonic buffered eyewash solution helps to clear eyes of foreign material such as dust, pollen, and chemicals, and to relieve itching and burning of the eyes and skin. The plastic squeeze bottle with tear-off top helps create a fluid stream to direct the solution into the affected areas.
For solution refills, please see items 76049-014 (16 oz.) and 76049-016 (32 oz.).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty