Would you like to log in again?
VWR® First Aid Metal Cabinets, ANSI A+, Type I & II, with Medications
Supplier: VWR
VWR®
76049-056 76049-060
76049-060CS 961.95 USD InStock
76049-060 76049-056
VWR® First Aid Metal Cabinets, ANSI A+, Type I & II, with Medications
First Aid Supplies First Aid Cabinets
These steel first aid cabinets meet ANSI standards and are designed for worksites and companies with 75+ employees.
  • Solid steel case, with carrying handle
  • Wall-mountable
  • Available in a 2-shelf or 3-shelf configuation
  • Available with 347 or 669 pieces
This solid steel case comes with a carrying handle and is wall-mountable.
Certifications: ANSI/ISEA Z308.1-2015. The 2-shelf cabinet exceeds the 2015 Class B requirements, and is sufficient to treat 75 to 100 people. The 3-shelf cabinet exceeds the 2015 Class A requirements, and is sufficient to treat 100 or more people.
Ordering information: The 2-Shelf Cabinet contains (1) fabric bandages, 1×3, 50ct.; (1) antibiotic ointment packets, 10ct.; (1) antiseptic spray, 4oz.; (1) CPR mask with one-way valve; (1) burn dressing, 4×4"; (1) burn spray, 4 oz.; (1) cold compress, 4×5"; (1) 4 sterile eye pads and 4 strips; (1) eyewash, 1 oz.; (2) conforming gauze rolls, 2"; (1) scissors; (1) triangular Sling/Bandage, 40×40×56"; (1) first aid guide; (1) tweezers; (1) plastic bandages, ¾×3", 100ct.; (1) hand sanitizer packets, 0.9 g, 10ct.; (2) gauze dressing pads, 3×3", 2ct.; (1) first aid tape, 1/2" ×5 yds.; (1) knuckle fabric bandages, 8ct.; (1) fingertip fabric bandages, 10ct.; (2) alcohol wipes, 10ct.; (1) aspirin tablets, 50ct.; (1) non-aspirin Tablets, 50ct.; (1) hema-fle× 5×9" trauma pad in 4×4" gauze; (2) nitrile gloves; (1) trauma pad, 5×9"; (2) nitrile e×am gloves.
The 3-shelf cabinet contains (1) fabric bandages, 1×3", 100ct.; (1) First aid tape roll, ½"×5 yds.; (1) Nitrile e×am gloves, 10ct.; (1) triple antibiotic ointment packets, 25ct.; (1) antiseptic spray, 4 oz.; (1) CPR mask with one-way valve; (1) Burn dressing, 4×4"; (1) burn spray, 4 oz.; (1) cold compress, 6×9"; (1) 4 sterile eye Pads and 4 strips; (1) eyewash, 4 oz.; (1) hand sanitizer packets, 0.9 g, 10ct.; (1) conforming gauze roll, 2"; (1) scissors; (1) gauze dressing pads, 3×3", 10ct.; (2) triangular sling/bandages, 40×40×56"; (1) first aid guide; (1) tweezers; (1) fabric bandages, 2×3", 10ct.; (1) fabric knuckle and fingertip bandage, 50ct.; (5) gauze dressing pads, 2×2", 2ct.; (5) gauze dressing pads, 4×4", 2ct.; (1) alcohol wipes, 100ct.; (1) non-aspirin tablets, 100ct.; (1) cotton tipped applicators, 3", 100ct.; (1) elastic bandage, 4" × 5 yds.; (1) hema-flex 5×9" trauma pad in 4×4" gauze; (2) nitrile gloves; (1) trauma pad, 5×9"; (1) antacid tablets, 100ct.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty