Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
SiliaFlash® Irregular Silica Gels, SiliCycle Inc
Supplier: SiliCycle
R12030B-1KG R10160H-1KG R10180D-1KG R10140H-1KG R12030B-32KG R10170B-1KG R10180H-1KG R10157B-1KG R10180T-1KG R10160B-1KG R10170M-1KG R10140D-1KG R10130M-1KG R10157B-1KG R10157D-1KG R10030B-25KG R10130D-1KG R10170H-1KG R10140B-10KG R10023B-1KG R10180B-1KG R10140D-1KG R10140B-5KG R10180B-1KG R10140M-1KG R10170D-1KG R10160H-1KG R12030B-10KG R10150B-1KG R10030B-1KG R10160B-1KG R10040B-1KG R10140B-25KG R10180P-1KG R10150H-1KG R10170B-1KG R10072H-1KG R10140B-5KG R10180T-1KG R10180H-1KG R10130M-1KG R10030B-1KG R10023B-1KG R10170H-1KG R10130D-1KG R10040B-1KG R10140B-1KG R10170M-1KG R10140B-25KG R12030B-5KG R10180P-1KG R10130H-1KG R10130H-1KG R10157H-1KG R10050B-1KG R10140A-1KG R10140M-1KG R10150H-1KG R10157D-1KG R10140B-10KG R10140H-1KG R10140A-1KG R10030B-5KG R10170D-1KG R10157H-1KG R10072H-1KG R12030B-1KG R12030B-25KG R10030B-10KG R10137L-1KG R10137L-1KG R10140B-1KG R10180D-1KG
76049-284EA 1001.91 USD InStock
76049-284 CA75873-738 75873-726 76049-288 CA75873-736 76049-286 75873-728 CA75873-734 75873-722 76049-248 CA75873-698 CA75873-732 CA75873-740 75873-730 75873-696 75873-732 75873-698 76049-290 76049-250 76049-294 CA75873-728 76049-292 75873-716 CA75873-726 75873-718 CA75873-722 75873-712 75873-756 CA75873-720 75873-714 CA75873-696 CA75873-730 75873-720 75873-682 76122-202 75873-684 CA76049-270 CA75873-718 CA75873-716 75873-704 75873-748 CA75873-714 75873-706 75873-744 CA75873-712 CA75873-756 75873-700 CA75873-754 75873-702 75873-746 75873-752 75873-754 CA75873-684 CA75873-682 75873-750 CA75873-706 76049-270 75873-738 CA75873-704 CA76049-286 CA75873-748 CA75873-746 CA75873-702 CA76049-284 CA75873-744 CA75873-700 75873-734 CA75873-742 75873-736 CA75873-752 75873-740 CA75873-750 75873-742
SiliaFlash® Irregular Silica Gels, SiliCycle Inc
Chromatography Sorbents Flash Chromatography Sorbents
SiliaFlash® products are ideal for both analytical and preparative chromatography, from laboratory to pilot-plant processes and production scales.
  • High purity silica gels
  • Lowest level of fines on the market
  • Exceptional narrow particle and pore size distributions
  • Batch-to-batch, year-to-year consistency
  • Neutral pH
  • Low metal content and controlled water content
With pore diameters ranging from 30 to 1,000 Ångström (Å) and particle sizes up to 1,000 microns (μm), SiliCycle offers products to meet all your requirements. We offer one of the most reliable portfolios for flash and gravity grades for low to medium-high pressure. Our silica gels are ideal for preparative chromatography, from laboratory to pilot-plant processes and production scale.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty