Would you like to log in again?
ROD STAND STAINLESS STEEL 10MM TITRATOR
Supplier: XYLEM/YSI
Stand rod 10 mm^ Stand rod, Material: Stainless steel, For TitroLine* KF trace, TitroLine* 7500 KF trace, TitroLine* 7500 KF, TitroLine* 5000 and TITRONIC* 300, Size: 10 mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ROD STAND STAINLESS STEEL 10MM TITRATOR 76049-466 285216560 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty