Would you like to log in again?
MegaMan® MM-211-120-BL Heavy-Duty Nitrile Single-Use Gloves, SW Safety Solutions
Supplier: SW Safety Solutions
N772353 N772356 N772352 N772351
76051-158BX 51.56 USD InStock
76051-158 76051-168 76051-162 76051-160
MegaMan® MM-211-120-BL Heavy-Duty Nitrile Single-Use Gloves, SW Safety Solutions
Gloves
PowerForm® gloves works with complete confidence knowing that this heavy-duty extended cuff glove has wearers covered, offering excellent grip and optimal chemical resistance.
  • Fully textured surface for superior wet and oily grip
  • Heavy-duty chemical resistance
  • Touch-screen compatible
  • Powder-free to prevent contamination
  • Clean processing technology helps prevent skin irritation and workplace contamination
  • Optimal thickness for durability and tactile sensitivity
MegaMan® pushes the boundaries of single-use hand protection performance, with new levels of abrasion resistance, sweat management, and comfort that deliver in exceedingly severe environments. Reach for MegaMan® on the toughest jobs, when protection matters most.
Loaded with features that add to this glove's performance capability, this glove line is designed and built to the very highest standards. Featuring 14 mil fingertips combined with a 12 mil palm thickness, this single-use glove also offers a full 12" length for unmatched splash protection and the ability to provide additional coverage to lab sleeves.
For decades SW® has been the premium manufacturer of single-use performance products. With a relentless focus on new manufacturing technologies and materials that drive innovation, SW® single-use styles continuously push the boundaries of quality and worker-enhancing performance that fuels business success. The SW® difference: 100% air inspected, 75% less residue, consistent tip to cuff.
Certifications: WBENC Women Owned; NCMSDC Minority Business; Small Business Enterprise; EAC TP TC 019/2011; CE 0493; MegaMan® is Dermatalogically Approved by the Skin Health Alliance (SHA); Meets stringent standards that ensure skin safety.
Ordering information: Glove sizes are as follows; XS (5.5 to 6), S (6.5 to 7), M (7.5 to 8), L (8.5 to 9), XL (9.5 to 10), XXL (10.5 to 11).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty