Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Upright Freezer Racks for 2" Boxes
Supplier: VWR
VWR®
UF-462 UF-332 UF-442 UFD-452-ST UF-362 UF-452 UF-542 UF-342 UF-552 UF-472 UF-232 UF-242 UF-432
76051-424EA 309.14 USD InStock
76051-424 76051-422 76530-432 76027-814 76051-408 76051-418 76027-800 76027-810 76051-406 76027-604 76027-812 76027-796 76027-798
VWR® Upright Freezer Racks for 2" Boxes
Racks Cryogenic Box Racks
Organize freezer samples.
  • Organize and find samples quickly
  • Space-saving design for specific freezer requirements
  • Security locking devices available
Racks are designed for use with standard 2" cardboard, plastic, or metal boxes. Racks are made from corrosion-resistant stainless steel. Security lock devices are also available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty