Would you like to log in again?
VWR® Freezer Drawer Racks
Supplier: VWR
VWR®
UFD-532 UFD-242 UFD-472 UFD-262 UFD-362 UFD-352 UFD-252 UFD-562 UFD-432
76051-546EA 400.48 USD InStock
76051-546 76051-534 76051-544 76051-532 76051-542 76051-530 76051-540 76051-538 76051-536
VWR® Freezer Drawer Racks
Racks Cryogenic Box Racks
Drawer rack for 2" boxes.
  • Stainless steel
  • Double welds
  • Smoothed edges
  • Lifetime warranty
These freezer racks are made to the highest levels of quality with pharmaceutical grade stainless steel, double welds, smoothed edges and an unconditional lifetime warranty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty