Would you like to log in again?
VWR® Sliding Tray Racks
Supplier: VWR
VWR®
UFS-432 UFS-422 UFS-442 UFS-552 UFS-352 UFS-542 UFS-452 UFS-522 UFS-342 UFS-532
76051-556EA 169.92 USD InStock
76051-556 76051-566 76051-554 76051-564 76051-552 76051-562 76051-550 76051-560 76051-548 76051-558
VWR® Sliding Tray Racks
Racks Cryogenic Box Racks
Sliding tray rack for 2" boxes.
  • Stainless steel
  • Double welds
  • Smoothed edges
  • Lifetime warranty
These freezer racks are made to the highest levels of quality with pharmaceutical grade stainless steel, double welds, smoothed edges and an unconditional lifetime warranty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty