Would you like to log in again?
VWR® Vertical Racks with Spring Lock
Supplier: VWR
VWR®
MCF-13-2-S MCF-11-2-S MCF-10-2-S MCF-9-2-S MCF-12-2-S MCF-14-2-S MCF-15-2-S
76051-588EA 78.56 USD InStock
76051-588 76051-586 76051-584 76051-594 76051-582 76051-592 76051-590
VWR® Vertical Racks with Spring Lock
Racks Cryogenic Box Racks
Vertical racks for mini 2" boxes with spring lock.
  • Stainless steel
  • Double welds
  • Smoothed edges
  • Lifetime warranty
Freezer Racks are made to the highest levels of quality with pharmaceutical grade stainless steel, double welds, smoothed edges and an unconditional lifetime warranty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty