Would you like to log in again?
VWR® Upright Sliding Tray Racks
Supplier: VWR
VWR®
UFS-543 UFS-423 UFS-333 UFS-453 UFS-443 UFS-523 UFS-533 UFS-343 UFS-433
76051-644EA 166.32 USD InStock
76051-644 76051-654 76051-642 76051-652 76051-640 76051-650 76051-638 76051-648 76051-646
VWR® Upright Sliding Tray Racks
Racks Cryogenic Box Racks
Upright sliding tray racks for 3" boxes.
  • Stainless steel
  • Double welds
  • Smoothed edges
  • Lifetime warranty
Freezer Racks are made to the highest levels of quality with pharmaceutical grade stainless steel, double welds, smoothed edges and an unconditional lifetime warranty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty