Would you like to log in again?
VWR® Vertical Racks
Supplier: VWR
VWR®
CF-6-3 CF-8-3 CF-4-3 CF-5-3 CF-7-3
76051-656EA 42.93 USD InStock
76051-656 76051-664 76051-662 76051-660 76051-658
VWR® Vertical Racks
Racks Cryogenic Box Racks
Vertical racks for 3" boxes.
  • Stainless steel
  • Double welds
  • Smoothed edges
  • Lifetime warranty
Freezer Racks are made to the highest levels of quality with pharmaceutical grade stainless steel, double welds, smoothed edges and an unconditional lifetime warranty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty