Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
VWR® Vertical Freezer Racks
Supplier: Avantor
VWR®
MCF-6-2 MCF-8-2 MCF-4-2 MCF-5-2 MCF-9-2 MCF-11-2 MCF-10-2 MCF-7-2
76051-678EA 24.65 USD InStock
76051-678 76051-688 76051-686 76051-684 76051-682 76051-692 76051-680 76051-690
VWR® Vertical Freezer Racks
Racks Cryogenic Box Racks
Vertical freezer racks for 2" mini boxes.
  • Stainless steel
  • Double welds
  • Smoothed edges
  • Lifetime warranty
Freezer Racks are made to the highest levels of quality with pharmaceutical grade stainless steel, double welds, smoothed edges and an unconditional lifetime warranty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty