Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
VWR® Vertical Freezer Racks
Supplier: Avantor
VWR®
MCF-8-3 MCF-7-3 MCF-4-3 MCF-5-3 MCF-6-3 MCF-9-3 MCF-10-3
76051-710EA 31.98 USD InStock
76051-710 76051-720 76051-708 76051-718 76051-716 76051-714 76051-712
VWR® Vertical Freezer Racks
Racks Cryogenic Box Racks
Vertical freezer racks for 3" mini boxes.
  • Stainless steel
  • Double welds
  • Smoothed edges
  • Lifetime warranty
Freezer Racks are made to the highest levels of quality with pharmaceutical grade stainless steel, double welds, smoothed edges and an unconditional lifetime warranty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty