Would you like to log in again?
VWR MIXED STORAGERACK FOR 12X2&8X3 BOX
Supplier: VWR International
VWR* Upright Freezer Rack 550 x 159 x 140 mm^ VWR* Upright Freezer Rack, Material: Stainless Steel, Box capacity: 12x2 in box, 8 x3 in box, 4x2 in +4 x3 in configuration, Outside Dimension: 550 x 159 x 140 mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR MIXED STORAGERACK FOR 12X2&8X3 BOX 76051-778 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty