Would you like to log in again?
VWR DRAWERRACK7X6 FOR 96D-WELL PLATES
Supplier: VWR International
VWR* Upright Stainless Steel Freezer Drawer Rack 635 x 307 x 138 mm^ VWR* Upright Stainless Steel Freezer Drawer Rack, Box Capacity: 7 deep & 6 high configuration(42), for 96 deep-well microtiter plates/Matrix/ Fluidx/Micronic plates, Outside Dimension: 635 x 307 x 138 mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR DRAWERRACK7X6 FOR 96D-WELL PLATES 76051-920 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty