Would you like to log in again?
VWR DRAWERRACK523X114X3 FOR 70MM BOTTLES
Supplier: VWR International
VWR* Upright Freezer Drawer Rack 559 x 231 x 140 mm^ VWR* Upright Freezer Drawer Rack, Material: Stainless Steel, Box capacity: 523x114mm & 523x114m & 523x114mm (3 drawer), For storage H less than or equal to 70mm bottle, Outside Dimension: 559 x 231 x 140 mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR DRAWERRACK523X114X3 FOR 70MM BOTTLES 76052-084 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty