Would you like to log in again?
VWR DRAWERRACK FOR 170X4/4.5/6/7ML TUBES
Supplier: VWR International
VWR* Upright Freezer Drawer Rack 419 x 228 x 140 mm^ VWR* Upright Freezer Drawer Rack, Material: Stainless Steel, Box capacity: 170tubes, For 4/4.5/6/ 7ml blood sample tubes (diameter less than or equal to 1/2 inch H less than or equal to 4-3/16 inch), Outside Dimension: 419 x 228 x 140 mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR DRAWERRACK FOR 170X4/4.5/6/7ML TUBES 76052-098 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty