Would you like to log in again?
VWR UPRIGHT RACK6X3FORH101.5~107.8PLATES
Supplier: VWR International
VWR* Upright Freezer Rack 540 x 336 x 138 mm^ VWR* Upright Freezer Rack, Material: Stainless Steel, Box capacity: 6 deep & 3 high configuration( 18), For Fluidx/Micronic plates( L127.7 xW85.7 xH88.5mm), Outside Dimension: 540 x 336 x 138 mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR UPRIGHT RACK6X3FORH101.5~107.8PLATES 76052-166 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty