Would you like to log in again?
VWR DRAWERRACK5X2FOR H23.8~28.5PLATES
Supplier: VWR International
VWR* Upright Freezer Drawer Rack 454.6 x 136 x 138 mm^ VWR* Upright Freezer Drawer Rack, Material: Stainless Steel, Box capacity: 5 deep & 2 high x 2 configuration( 20), For Matrix/ Fluidx/Micronic/Abgene 96- deep-Well Microtiter plates, Outside Dimension: 454.6 x 136 x 138 mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR DRAWERRACK5X2FOR H23.8~28.5PLATES 76052-188 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty