Would you like to log in again?
VWR VERTICALRACK1X15 FOR 96D-WELL PLATES
Supplier: VWR International
VWR* Vertical Rack 694 x 92 x 140 mm^ VWR* Vertical Rack, Material: Stainless Steel, Box capacity: 15plates, For Chest Freezers, For 96 deep-well microtiter plates/Matrix/ Fluidx/Micronic plates, Outside Dimension: 694 x 92 x 140 mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR VERTICALRACK1X15 FOR 96D-WELL PLATES 76052-208 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty