Would you like to log in again?
VWR UPRIGHT RACK5X5FOR 96-W PCR BOXES
Supplier: VWR International
VWR* Upright Freezer Rack 466 x 190 x 140 mm^ VWR* Upright Freezer Rack, Material: Stainless Steel, Box capacity: 5 deep & 5 high configuration(25), For 0.2ml tube 96-well PCR boxes, Outside Dimension: 466 x 190 x 140 mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR UPRIGHT RACK5X5FOR 96-W PCR BOXES 76052-380 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty