Would you like to log in again?
VWR UPRIGHT RACK3X5FOR H55.9~61.8 PLATES
Supplier: VWR International
VWR* Upright Freezer Rack 271.2 x 317 x 138 mm^ VWR* Upright Freezer Rack, Material: Stainless Steel, Box capacity: 3 deep & 5 high configuration(15), For Matrix/ Fluidx/Micronic/Abgene plates, Outside Dimension: 271.2 x 317 x 138 mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR UPRIGHT RACK3X5FOR H55.9~61.8 PLATES 76052-444 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty