Would you like to log in again?
VWR DRAWER RACK4X3 FOR 3.75 BOXES
Supplier: VWR International
VWR* Upright Stainless Steel Freezer Drawer Rack 559x318x140mm^ VWR* Upright Stainless Steel Freezer Drawer Rack, 4 deep /3high configuration 12 x3.75in high standard box capacity, Dimension: 559x318x140mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR DRAWER RACK4X3 FOR 3.75 BOXES 76052-640 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty