Would you like to log in again?
VWR VERTICAL 1X9 FOR 96-WELL PLATES
Supplier: VWR International
VWR* Vertical Stainless Steel Rack 506x92x140mm^ VWR* Vertical Stainless Steel Racks For Chest Freezers, for 96-well microtube boxes//Matrix/ Fluidx/Micronic/Abgene plates( L127.7 xW85.7 xH(17.3x3;49.1 approx 54mm)), configuration 9microtube boxes capacity, Dimension: 506x92x140mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR VERTICAL 1X9 FOR 96-WELL PLATES 76052-670 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty