Would you like to log in again?
VWR VERTICALRACK4X2 FOR 50C 0.5ML BOXES
Supplier: VWR International
VWR* Vertical Stainless Steel Rack 228x140x140mm^ VWR* Vertical Stainless Steel Racks For Chest Freezers, for 50-cell 0.5ml storage boxes, configuration 8boxes capacity, Dimension: 228x140x140mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR VERTICALRACK4X2 FOR 50C 0.5ML BOXES 76052-720 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty