Would you like to log in again?
VWR DRAWERRACK5X6FOR H23.8~28.5PLATES
Supplier: VWR International
VWR* Upright Stainless Steel Freezer Drawer Rack^ VWR* Upright Stainless Steel Freezer Drawer Rack, for Matrix/ Fluidx/Micronic/Abgene plates( L127.7 x W85.7 x H23.8 approx 28.5mm ), 5deep /6high configuration 60plates capacity, Dimension:
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR DRAWERRACK5X6FOR H23.8~28.5PLATES 76052-788 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty