Would you like to log in again?
VWR DRAWERRACK4X3FOR H31.7~38.7PLATES
Supplier: VWR International
VWR* Upright Stainless Steel Freezer Drawer Rack 361x142x138mm^ VWR* Upright Stainless Steel Freezer Drawer Rack, for Fluidx/Micronic/Abgene plates( L127.7 xW85.7 xH(31.7 approx 38.7mm)), 4deep /3high configuration 12plates capacity, Dimension: 361x142x138mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR DRAWERRACK4X3FOR H31.7~38.7PLATES 76052-800 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty