Would you like to log in again?
VWR MODIFIABLE RACK FOR 15X2&10X3 BOXES
Supplier: VWR International
VWR* Upright Stainless Steel Freezer Modifiable Rack 681x168.8x140mm^ VWR* Upright Stainless Steel Freezer Modifiable Rack, configuration 15x2in box, 10 x3in box, 5x2in + 5 x3in box capacity, Dimension: 681x168.8x140mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR MODIFIABLE RACK FOR 15X2&10X3 BOXES 76053-078 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty