Would you like to log in again?
SW® TrueForm® TF-95LG Light Green 4 mil Nitrile Exam EnerGel® Rejuvenating Gloves
Supplier: SW Safety Solutions
SW® TrueForm®
N128402 N128401 N128405 N128404 N128403
76053-100BX 21.2 USD InStock
76053-100 76053-098 76053-096 76053-094 76050-932
SW® TrueForm® TF-95LG Light Green 4 mil Nitrile Exam EnerGel® Rejuvenating Gloves
Gloves
Featuring an EnerGel™ inner coating, these gloves soothe and moisturize dry hands without sacrificing dexterity.
  • EnerGel® hand conditioning technology hydrates and soothes skin
  • EcoTek® sustainable technology
  • Optimal thickness for durability and tactile sensitivity
  • Fully textured surface for excellent wet and dry grip
  • Touch-screen compatible
  • Tear-resistant beaded cuff eases donning
  • SHA dermatologically approved
  • 1.5 AQL exceeds laboratory standards
Featuring an EnerGel® inner coating, these gloves soothe and moisturize dry hands without sacrificing dexterity. EcoTek® gloves tested to ASTM D5526-12 standard test method for determining anaerobic biodegradation of plastic materials under accelerated landfill conditions. Results are 92.6% biodegradation in 60% solids landfill at 945 days. Future results cannot be predicted/extrapolated. For additional information on the test reports, contact cs@swssglobal.com.
Certifications: WBENC Women Owned; NCMSDC Minority Business; Small Business Enterprise; SHA Dermatologically Approved; Meets stringent standards that ensure skin safety; EAC TP TC 019/2011; CE 0493; CE EN 455.
Ordering information: Glove sizes are as follows; XS (5.5 to 6), S (6.5 to 7), M (7.5 to 8), L (8.5 to 9), XL (9.5 to 10).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty