Would you like to log in again?
VWR UPRIGHT RACK4X14 FOR 96/384-W PLATES
Supplier: VWR International
VWR* Upright Stainless Steel Freezer Rack 374x233x140mm^ VWR* Upright Stainless Steel Freezer Rack, for 96/384-well microtiter plates configuration 56 plates w/ lids or 64 plates w/o lids capacity, Dimension: 386 x 140 x 233mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR UPRIGHT RACK4X14 FOR 96/384-W PLATES 76053-120 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty