Would you like to log in again?
VWR VERTICALRACK1X14 - 25-P SLIDE BOXES
Supplier: VWR International
VWR* Vertical Stainless Steel Rack 540x152x97mm^ VWR* Vertical Stainless Steel Racks For Chest Freezers, for 25-place slide boxes, configuration 14boxes capacity, Dimension: 540x152x97mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR VERTICALRACK1X14 - 25-P SLIDE BOXES 76053-170 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty