Would you like to log in again?
VWR BIN ORGANIZER WITH LID546X121X134
Supplier: VWR International
VWR* Bin organizer with lid 546x121x134mm^ VWR* Bin organizer with lid Excellent for organizing -20* Freezers/cold rooms, Constructed of corrosion resistant stainless steel, Dimension: 554 x 120 x 138 mm, Drawer Area: 546 x 121 x 134 mm, Shipping Weight: 3.53lb
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR BIN ORGANIZER WITH LID546X121X134 76053-250 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty