Would you like to log in again?
VWR BIN ORGANIZER WITH LID666X224X134
Supplier: VWR International
VWR* Bin organizer with lid 666x224x134mm^ VWR* Bin organizer with lid Excellent for organizing -20* Freezers/cold rooms, Constructed of corrosion resistant stainless steel, Dimension: 674 x 225 x 138 mm, Drawer Area: 666 x 224 x 134 mm, Shipping Weight: 4.98lbs
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR BIN ORGANIZER WITH LID666X224X134 76053-264 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty