Would you like to log in again?
VWR DRAWERRACK BIN ORGANIZE406X119X274X2
Supplier: VWR International
VWR* Drawer Rack bin organizer 406x119x274x2mm^ VWR* Drawer Rack bin organizer Excellent for organizing -20* Freezers/cold rooms, Constructed of corrosion resistant stainless steel, Dimension: 419 x 252 x 284 mm, Drawer Area: 416 x 124 x 282 mm, Shipping Weight: 13.87lbs
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR DRAWERRACK BIN ORGANIZE406X119X274X2 76053-312 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty