Would you like to log in again?
VWR SECURITY LOCK FOR H670 RACK
Supplier: VWR International
VWR* Security Lock Devices 660mm, Height: 670mm^ VWR* Security Lock Devices for Chest Freezer/Liquid Nitrogen Tank Racks FOR H670 RACK, Material: Stainless steel, Dimension: 660mm, Height: 670mm, Applicable to Rack: CF-12-2, CF-12-2-S, CFA-12-2
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR SECURITY LOCK FOR H670 RACK 76053-364 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty