Would you like to log in again?
Filter Degasser Kit
Supplier: MicroSolv
58800-47-KT-2L 58800-47-KT 58800-48-KT-2L
89220-030EA 698.75 USD InStock
89220-030 76057-642 76057-636
Filter Degasser Kit
Chromatography Supplies Chromatography Spare Parts
This borosilicate glass, Mobile Phase Filtration Assembly is great for filtering and degassing HPLC solvents for use as mobile phases.
The unit’s design of the fritted glass support for the membrane also prevents contamination of your vacuum line or source which is separate from this unit. The funnels are graduated and will holds 300 to 500 ml of mobile phase. The reservoir flasks holds 1000 and 2000 ml and the narrow mouth opening minimizes out-gassing.
Use any disposable, 47 mm membrane disc that is compatible with your mobile phase.
Ordering information: The Filter Degasser Assembly is sold as a complete kit including the blue metal clamp. Replacements parts are available upon request. Membranes are sold separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty