Would you like to log in again?
Pico™ and Fresco™ Microcentrifuges, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Pico™ Fresco™
75002554 75002416 75772411 75002401 75772492 75002411 75002426 75002402 75772478 75002421 75772477 75002477 75002556
76057-714EA 8656.09 USD InStock
76057-714 97007-114 97007-116 76057-718 76057-720 89201-372 97007-110 97000-048 97007-112 76057-712 89201-374 76057-656 76057-722
Pico™ and Fresco™ Microcentrifuges, Thermo Scientific
Centrifuges Benchtop Centrifuges
Benchtop microcentrifuges are ideal for a broad range of bioresearch separation tasks in the microliter sample range, such as nucleic acid separation, protein isolation, and reaction set-up
Microcentrifuges are lightweight with a small footprint and feature a frequency-controlled, brushless induction drive motor for quiet, reliable, vibration-free operation. Steel armored rotor chamber interlocks with lid to provide uncompromised user safety. Standard and hematocrit 24-place rotors come with CAMR-certified ClickSeal™ biocontainment lid. Dual 18-place rotor comes with screw-on lid. Both microcentrifuges feature twelve second acceleration and deceleration for faster sample processing. Easy-to-read LED displays indicate speed and run time (plus temperature on Fresco model). Microprocessor controller ensures precise, reproducible results. Flat membrane keypad simplifies operation and makes cleaning easy. Parameter settings are maintained in memory until new data is entered — even in the event of a power outage. Rotor materials are autoclavable and designed to withstand vigorous cleaning.
Pico™ model centrifuges are ventilated. Fresco™ model centrifuges are refrigerated with an environmentally safe CFC-free refrigerant, and feature stand-still cooling that chills samples prior to use.
Certifications: CSA certified, CE Marked, IVD compliant, Certified Bio-containment.
Ordering information: Supplied with a two-year parts and labor warranty with one additional year redemption. Microcentrifuges are available with either a 24x1.5–2.0mL rotor with a ClickSeal biocontainment lid, or a dual 18-place rotor with screw-on lid, or both. Hematocrit rotor may be purchased separately and swapped with standard rotor. For other rotor options, contact your Avantor sales representative. Adapters must also be purchased separately.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICES
Services: Thermo Scientific provides several service options such as Extended Warranty, Extended Warranty plus on-site Annual Preventative Maintenance, and Total Care. To learn more about these services please search "Thermo Centrifuge Technical Service" in the website search field.
In addition, VWRCATALYST™ can provide an array of services such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWRCATALYST@vwr.com for further details.
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty