Would you like to log in again?
Puritan® PurSafe® DNA/RNA Preservative, Puritan Medical Products
Supplier: Puritan Medical Products
Puritan®
VL1201SAFE-H VL1201SAFE VL1202SAFE VL1202SAFE-H VL1201SAFE VL1201SAFE-H VL1202SAFE-H VL1202SAFE
76057-734BX 328.82 USD InStock
76057-734 76057-624 76057-736 76057-626 CA76057-624 CA76057-734 CA76057-736 CA76057-626
Puritan® PurSafe® DNA/RNA Preservative, Puritan Medical Products
Vials
Intended for the stabilization and preservation of DNA and RNA of specimens to be analyzed by nucleic acids amplification tests.
  • Compatible with various sample types
  • Total microbial inactivation
  • Compatible with preservation of DNA/RNA from bacteria, viruses and tissues
  • HydraFlock® flocked swab maximizes sample collection and elution
  • For research use only
Simplify specimen collection and preservation for molecular research with PurSafe®.
Ordering information: Products are final sale.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty