Would you like to log in again?
Cyclic AMP TR-FRET assay kit, Columbia Biosciences
Supplier: Columbia Biosciences
AD-CAMP-100K AD-CAMP-1K AD-CAMP-20K
76076-702EA 15048.42 USD InStock
76076-702 76076-700 76076-698
Cyclic AMP TR-FRET assay kit, Columbia Biosciences
Assays Cellular Assays
Homogenous, TR-FRET based assay for detection of cAMP
  • Homogenous assay , mix and read
  • No wash steps
  • Eliminates compound interference
  • Allows for HTS assays to be performed in a miniaturized format.
  • TR-FRET format increases sensitivity
This kit is designed to to detect and quantitatively measure the amount of cyclic AMP (cAMP) present in cell based assay systems. A europium (Eu) labeled monoclonal anti-cAMP antibody (Donor) is paired with a fluorescently labeled cAMP analog (Acceptor), and in the absence of any endogenous cAMP, will produce a time-resolved energy transfer signal at 665 nm when excited at 340 nm. If cAMP accumulates in the well due to the activation of cellular adenylyl cyclase, this cAMP inhibits the formation of the labeled Donor/Acceptor complex thereby decreasing the amount of energy transfer signal produced and the overall drop in signal is inversely proportional to the amount of cAMP accumulated.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty