Would you like to log in again?
Q-SWAB™ Dry Swab with Transport Tube, Simport
Supplier: Simport
T307-10A T307-5A
76076-726CS 476.79 USD InStock
76076-726 76076-724
Q-SWAB™ Dry Swab with Transport Tube, Simport
Swabs Swabs for Sampling
Sterile specimen collection swabs.
  • Polypropylene tubes are translucent
  • Cap is equipped with a silicone washer
  • Non-graduated
The Q-Swab™ design is self-standing, but with round bottom. The polyester swab holds securely inside the screw cap, making it ideal for specimen collection and protection from contamination. The tube material make it easy to see all contents. The cap features with washer fitted inside to ensure a positive seal at any temperature.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty