Would you like to log in again?
Glass Microscope Slides, Azer Scientific
Supplier: Azer Scientific
2752510A 2752512A 2752511
76268-418CS 348.26 USD InStock
76268-418 76076-736 76076-734
Glass Microscope Slides, Azer Scientific
Slides Microscope Slides
General-purpose microscope slides offer quality and value for any laboratory.
  • Extra white soda-lime glass with 90 ° corners
  • Manufactured in Germany
Choose any one of these general-purpose microscope slides for clinical, environmental, industrial, or other routine microscopy work. Extra white soda-lime glass will not interfere with fluorescent staining procedures. Uniform 1.0 mm thickness makes these sturdy enough for field work.
Packaging: Slides are packaged 72 per box, 20 boxes per case.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty