Would you like to log in again?
Unifrost™ Adhesive Microscope Slides, Azer Scientific
Supplier: Azer Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
EMS500W+
76076-740CS 709.38 USD InStock
76076-740
Unifrost™ Adhesive Microscope Slides, Azer Scientific
Slides Adhesive Slides
These adhesive microscope slides are coated with Poly-L-Lysine.
  • Alternative to silane coated slides
  • Ideal for IHC procedures
  • Designed for the toughest, most difficult procedures
Permanent adhesive firmly anchors all specimen types to the slides.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty