Would you like to log in again?
Orion™ Star™ A123 Dissolved Oxygen Portable Meter, Thermo Scientific
Supplier: Thermo Scientific
Orion™ Star™
STARA1235 STARA1236 STARA1230
76076-750EA 2724.75 USD InStock
76076-750 89206-298 89206-300
Orion™ Star™ A123 Dissolved Oxygen Portable Meter, Thermo Scientific
Electrochemical Meters Oxygen Meters
Take simple dissolved oxygen and temperature measurements with the Orion™ Star™ A123 DO Portable Meter with easy operation for basic field testing.
  • Measure dissolved oxygen in percent saturation or mg/L with temperature using polarographic DO probes
  • Calibrate using water-saturated air or custom value (Winkler titration)
  • Automatically compensate DO readings using ATC temperature, manual salinity input, and manual barometric pressure
  • Simplified, easy-to-read display shows main measurement with temperature, stability indicator, and electrode condition icon
  • Easily recall calibration data for procedure checks
  • Non-volatile memory preserves 50 point internal data log and meter settings, even with loss of power
  • Three-year meter warranty
Take simple, routine dissolved oxygen and temperature measurements with the Orion™ Star™ A123 Dissolved Oxygen Portable Meter, an easy-to-use and budget-friendly instrument for basic dissolved oxygen analysis in the field.
These meters are a simple, economical solution for routine outdoor sample analysis. Featuring an emphasized measure key, selectable read modes, and measurement stability indicator, the basic Orion™ Star™ A123 dissolved oxygen meters are designed to deliver measurements users can trust at an affordable price. Flexible power options with AA batteries or optional universal power adapter.
Calibrate polarographic DO probes using water-saturated air or custom value using Winkler titration. Quickly navigate setup menus and ensure consistent calibrations using the simple keypad layout and on-screen text prompts. Use the included electrode stand to easily place sensors in samples and simplify storage. The IP67-rated housing is waterproof for rugged field operation.
Certifications: CE, TUV 3-1, FCC Class A.
Ordering information: For compatible DO probe, see Orion™ item 083005MD. For compatible buffer solutions, see Orion™ item 80513.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty