Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DURAN®PURE® Bottles, DWK Life Sciences

Supplier: DWK Life Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

818013608 891081306 818664409 818606301 818016305 818665405 818604403 818062402 818064403 891169109 818017301 818666307 818063604 818067306 818066301 891088679 818605408 818018606 818065408 818015403 898862802 818019105 818061406 818014407 818069101 818011401 818068602 818012406
76076-908CS 125.23 USD InStock
76076-908 76076-906 76076-904 76202-922 76202-924 76202-926 76202-928 76076-872 76076-894 76076-892 76076-870 76076-890 76076-912 76076-878 76076-910 76076-876 76076-898 76076-874 76076-896 76076-884 76076-882 76076-880 76202-920 76076-902 76076-868 76076-900 76076-888 76076-886
DURAN®PURE® Bottles, DWK Life Sciences
Bottles Glass Bottles
Intermediate biologic bottle system with comprehensive qualification and validation package.

  • USP compliant range of bottles from 25 ml to 20 L
  • Chemically inert
  • Wide usage temperature range
  • Excellent gas barrier
  • Low leachable/extractable profile
  • Biocompatibility

Bottles are made from Type 3.3 borosilicate glass. A protective dust cover is applied after the bottle is cooled on the production line.

DURAN PURE Premium Screw Caps are manufactured from fluoropolymer. The caps are colorant- and pigment-free and are suitable for most sterilization methods.

These validation ready sizes are scalable from pilot to production volumes, with the added durability to go from hot 'sterile' fills to cold storage. These bottles meet the documentation and production requirements of national and international pharmacopeias for primary packaging materials.

Certifications: DURAN PURE Bottles are supported by a comprehensive, easy to access, online validation document package and support services. The documentation package includes a certificate of lot analysis, Type III Drug Master File, and Qualification documentation.

Ordering information: Screw caps are sold separately.

Packaging: The bottles are packaged in shrinked-wrapped modules.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty